1H-Imidazole-4-carbonitrile 95%

Lieferant: Thermo Fisher Scientific
Gefahr

461360010 461360050
ACRO461360010EA 59.5 EUR
ACRO461360010 ACRO461360050
1H-Imidazole-4-carbonitrile 95%
1H-Imidazole-4-carbonitrile
Formel: C₄H₃N₃
Molecular Weight: 93.09 g/mol
Schmelzpunkt: 32…35 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233358
CAS-Nummer: 57090-88-7
UN: 3439
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 141 to 148 °C
Water ≤2.5 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR