Daptomycin

Lieferant: Thermo Fisher Scientific

461371000 461375000
ACRO461371000EA 144 EUR
ACRO461371000 ACRO461375000
Daptomycin
Daptomycin
Formel: C₇₂H₁₀₁N₁₇O₂₆
Lagertemperatur: Gefrierschrank
MDL: MFCD08282794
CAS-Nummer: 103060-53-3

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Solubility (0.5% in MeOH) Clear colorless to light yellow
HPLC ≥93 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR