Rosuvastatin sodium salt 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Rosuvastatin Sodium, Sodium (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulphonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate

461490010
ACRO461490010EA 165 EUR
ACRO461490010
Rosuvastatin sodium salt 98%
Rosuvastatin sodium salt
Formel: C₂₂H₂₇FN₃NaO₆S
Molecular Weight: 503,52 g/mol
Schmelzpunkt: 106…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD18783208
CAS-Nummer: 147098-18-8

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Water ≤2.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR