Stevioside
Lieferant: Thermo Scientific
Synonyme:
(2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate
461560250
ACRO461560250EA
76.8
EUR
ACRO461560250
Stevioside
Stevioside
Formel:
C₃₈H₆₀O₁₈ Molecular Weight: 804,88 g/mol Schmelzpunkt: 198 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00079561 CAS-Nummer: 57817-89-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤6 % |
HPLC | ≥75 % |
Specific optical rotation | -28° to -32° (20°C, 589 nm) (6 % in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...