Lenalidomide ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(4-Amino-1-oxo-1,3-dihydro-2H-isoindol-2-yl)piperidine-2,6-dione, (R,S)-Lenalidomide, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione

461590010
ACRO461590010EA 186 EUR
ACRO461590010
Lenalidomide ≥98%
Lenalidomide

  • Bioactive

Formel: C₁₃H₁₃N₃O₃
Molecular Weight: 259,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07772307
CAS-Nummer: 191732-72-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤3 %
HPLC ≥97.5 %
Residue after ignition ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR