Deferoxamine mesylate 95%

Lieferant: Thermo Fisher Scientific

Synonyme: DFOM

461770010
ACRO461770010EA 135 EUR
ACRO461770010
Deferoxamine mesylate 95%
Deferoxamine mesylate
Formel: C₂₆H₅₂N₆O₁₁S
Schmelzpunkt: 148…149 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00058605
CAS-Nummer: 138-14-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear to hazy Colorless to light brown-yellow
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR