Pentamidine isethionate salt 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pentamidine isethionate, 4,4'-(Pentane-1,5-diylbis(oxy))dibenzimidamide bis(2-hydroxyethanesulphonate)
461860010
ACRO461860010EA
158
EUR
ACRO461860010
Pentamidine isethionate salt 97%
Pentamidine isethionate salt
Formel:
C₁₉H₂₄N₄O₂·2C₂H₆O₄S Siedepunkt: 539,4 °C (760 mmHg) Schmelzpunkt: 188…194 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00079213 CAS-Nummer: 140-64-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 188 to 194 °C |
Loss on drying | ≤10 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...