Rifapentine 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-3-(((4-Cyclopentyl-1-piperazinyl)imino)methyl)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione

461900250 461901000
ACRO461900250EA 90.6 EUR
ACRO461900250 ACRO461901000
Rifapentine 95%
Rifapentine
Formel: C₄₇H₆₄N₄O₁₂
Schmelzpunkt: 179…180 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00866806
CAS-Nummer: 61379-65-5

Bestellen

Spezifikation Testergebnisse

Appearance Red to dark red Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR