PSI-7977

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-Isopropyl 2-(((S)-(((2R,3R,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-4-fluoro-3-hydroxy-4-methyltetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)amino)propanoate, Sofosbuvir

461932500
ACRO461932500EA 314 EUR
ACRO461932500
PSI-7977
PSI-7977

  • Bioactive

Formel: C₂₂H₂₉FN₃O₉P
Molecular Weight: 529,45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26523124
CAS-Nummer: 1190307-88-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR