Vecuronium bromide 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-[(2beta,3alpha,5alpha,16beta,17beta)-3,17-Bis(acetyloxy)-2-(1-piperidinyl)androstan-16-yl]-1-methylpiperidinium Bromide

461972500
ACRO461972500EA 104 EUR
ACRO461972500
Vecuronium bromide 98%
Vecuronium bromide
Formel: C₃₄H₅₇BrN₂O₄
Molecular Weight: 637.74 g/mol
Schmelzpunkt: 235°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00867762
CAS-Nummer: 50700-72-6
EINECS: 256-723-9
Merck Index: 13.10003

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤ 4% (105°C)
Titration with HClO4 97.5 to 102.5% (dry substance)
Specific optical rotation +30.5° to +35° (20°C, 589nm) (1% in 0.05M HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR