Raltegravir potassium salt
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
462160010
462162500
ACRO462160010EA
450
EUR
ACRO462160010
ACRO462162500
Raltegravir potassium salt
Formel:
C₂₀H₂₀FKN₆O₅ Schmelzpunkt: 155…157 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD12031642 CAS-Nummer: 871038-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...