Amisulpride

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-ethylsulphonyl-2-methoxybenzamide

462300010
ACRO462300010EA 271 EUR
ACRO462300010
Amisulpride
Amisulpride
Formel: C₁₇H₂₇N₃O₄S
Molecular Weight: 369,48 g/mol
Schmelzpunkt: 127 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00866691
CAS-Nummer: 71675-85-9

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR