Dasatinib monohydrate

Lieferant: Thermo Scientific
Gefahr

Synonyme: Dasatinib hydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide monohydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrate

462320010
ACRO462320010EA 190 EUR
ACRO462320010
Dasatinib monohydrate
Dasatinib monohydrate
Formel: C₂₂H₂₆ClN₇O₂S·H₂O
Molecular Weight: 506,02 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11046566
CAS-Nummer: 863127-77-9
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 2 to 4 %
HPLC ≥98.5 %
Residue after ignition ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR