(S)-Citalopram oxalate

Lieferant: Thermo Scientific

Synonyme: (S)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Oxalate

462340010
ACRO462340010EA 160 EUR
ACRO462340010
(S)-Citalopram oxalate
(S)-Citalopram oxalate

  • Bioactive

Formel: C₂₀H₂₁FN₂O
Molecular Weight: 414,43 g/mol
Schmelzpunkt: 146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06407826
CAS-Nummer: 219861-08-2

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Solubility (2 % in DMSO) Clear colorless solution
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR