Lapatinib
Lieferant: Thermo Scientific
Synonyme:
N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulphonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine, GW-572016
462360010
ACRO462360010EA
237
EUR
ACRO462360010
Lapatinib
Lapatinib
- Bioactive
Formel:
C₂₉H₂₆ClFN₄O₄S Molecular Weight: 581,1 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09264194 CAS-Nummer: 231277-92-2 |
Spezifikation Testergebnisse
Appearance | Light yellow to yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...