Ziprasidone hydrochloride monohydrate

Lieferant: Thermo Fisher Scientific

462430010 462432500
ACRO462430010EA 504 EUR
ACRO462430010 ACRO462432500
Ziprasidone hydrochloride monohydrate
Ziprasidone hydrochloride monohydrate

  • Bioactive

Formel: C₂₁H₂₁ClN₄OS·HCl·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00921885
CAS-Nummer: 138982-67-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light pink or light orange Powder
Infrared spectrum Conforms
Water 3.7 to 5.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR