Granisetron-hydrochlorid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide Hydrochloride
462480010
ACRO462480010EA
196
EUR
ACRO462480010
Granisetron-hydrochlorid ≥98%
Granisetron hydrochloride
- Bioactive
Formel:
C₁₈H₂₄N₄O Molecular Weight: 348,88 g/mol Schmelzpunkt: 284…290 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01747034 CAS-Nummer: 107007-99-8 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
Solubility | (1 % in water) Clear colorless solution |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...