Valdecoxib

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(5-methyl-3-phenylisoxazol-4-yl)benzenesulphonamide

462522500
ACRO462522500EA 131 EUR
ACRO462522500
Valdecoxib
Valdecoxib

  • Bioactive

Formel: C₁₆H₁₄N₂O₃S
Molecular Weight: 314,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00950568
CAS-Nummer: 181695-72-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 169 to 173 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR