Sunitinib malate

Lieferant: Thermo Fisher Scientific
Gefahr

462640010
ACRO462640010EA 88.2 EUR
ACRO462640010
Sunitinib malate
Sunitinib malate

  • Bioactive

Formel: C₂₂H₂₇FN₄O₂·C₄H₆O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD08282795
CAS-Nummer: 341031-54-7
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Solubility (1 % in DMSO) Clear yellow to yellow-orange solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR