Cyproheptadine hydrochloride sesquihydrate

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyproheptadine hydrochloride, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine hydrochloride

462670050
ACRO462670050EA 77.5 EUR
ACRO462670050
Cyproheptadine hydrochloride sesquihydrate
Cyproheptadine hydrochloride sesquihydrate

  • Bioactive

Formel: C₂₁H₂₁N·HCl·11/2H₂O
Molecular Weight: 350,88 g/mol
Schmelzpunkt: 165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012538
CAS-Nummer: 41354-29-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Water 7.0 to 9.0 %
Solubility (5 % in ethanol) clear colorless to light yellow
Titration with HClO4 98.5 to 100.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR