Loxapine succinate

Lieferant: Thermo Scientific

462690010
ACRO462690010EA 230 EUR
ACRO462690010
Loxapine succinate
Loxapine succinate

  • Bioactive

Formel: C₁₈H₁₈ClN₃O·C₄H₆O₄
Schmelzpunkt: 151…154 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00069298
CAS-Nummer: 27833-64-3

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Loss on drying ≤1 % (105 °C)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR