Eflornithine hydrochloride monohydrate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Eflornithine hydrochloride hydrate, DL-alpha-Difluoromethylornithine Hydrochloride, 2,5-Diamino-2-(difluoromethyl)pentanoic acid hydrochloride hydrate

462700010 462700050 462700250
ACRO462700010EA 49 EUR
ACRO462700010 ACRO462700050 ACRO462700250
Eflornithine hydrochloride monohydrate
Eflornithine hydrochloride monohydrate

  • Bioactive

Formel: C₆H₁₅ClF₂N₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD00242734
CAS-Nummer: 96020-91-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water 6 to 9 %
Titration with HClO4 ≥97.5 % (dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR