Tiamulin fumarate

Lieferant: Thermo Scientific
Achtung

462820010
ACRO462820010EA 161 EUR
ACRO462820010
Tiamulin fumarate
Tiamulin fumarate

  • Bioactive

Formel: C₃₂H₅₁NO₈S
Siedepunkt: 563 °C (760 mmHg)
Schmelzpunkt: 147…148 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00145407
CAS-Nummer: 55297-96-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 152 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Titration with NaOH 18.5 to 19.5 % (Fumaric acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR