Zanamivir monohydrate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2R,3R,4S)-3-Acetamido-4-guanidino-2-((1R,2R)-1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid hydrate

462960010
ACRO462960010EA 344 EUR
ACRO462960010
Zanamivir monohydrate
Zanamivir monohydrate

  • Bioactive

Formel: C₁₂H₂₀N₄O₇·H₂O
Molecular Weight: 350.33 g/mol
Schmelzpunkt: 256 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00866966
CAS-Nummer: 1260601-68-0

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying 4 % to 9 % (1 g, 105°C) (vacuum)
HPLC ≥97.5 %
Specific optical rotation (20°C, 589 nm) (on dry substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR