Palonosetron hydrochloride

Lieferant: Thermo Scientific

Synonyme: (S)-2-((S)-Quinuclidin-3-yl)-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride

462990010
ACRO462990010EA 369 EUR
ACRO462990010
Palonosetron hydrochloride
Palonosetron hydrochloride

  • Bioactive

Formel: C₁₉H₂₄N₂O
Molecular Weight: 332,87 g/mol
Schmelzpunkt: 307 °C (Zersetzung)
Lagertemperatur: Gefrierschrank
MDL: MFCD07370072
CAS-Nummer: 135729-62-3

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (80°C, 3 h) (vacuum)
HPLC ≥98.5 %
Identification Passes test for Chlorides
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR