(+)-Catechin monohydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
463020010
463020050
ACRO463020010EA
50.2
EUR
ACRO463020010
ACRO463020050
(+)-Catechin monohydrate
(+)-Catechin monohydrate
- Bioactive
Formel:
C₁₅H₁₄O₆·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075649 CAS-Nummer: 225937-10-0 |
Spezifikation Testergebnisse
Appearance | White to beige to light brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤8 % |
Solubility | (5 % in Ethanol, hot) Clear to hazy |
HPLC | ≥96.0 % |
Specific optical rotation | +24° to +28° (20°C, 589 nm) (1 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...