(+)-Catechin monohydrate

Lieferant: Thermo Fisher Scientific
Achtung

463020010 463020050
ACRO463020010EA 50.2 EUR
ACRO463020010 ACRO463020050
(+)-Catechin monohydrate
(+)-Catechin monohydrate

  • Bioactive

Formel: C₁₅H₁₄O₆·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00075649
CAS-Nummer: 225937-10-0

Bestellen

Spezifikation Testergebnisse

Appearance White to beige to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤8 %
Solubility (5 % in Ethanol, hot) Clear to hazy
HPLC ≥96.0 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR