Exemestane

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-Methyleneandrosta-1,4-diene-3,17-dione

463160010
ACRO463160010EA 90 EUR
ACRO463160010
Exemestane
Exemestane

  • Bioactive

Formel: C₂₀H₂₄O₂
Molecular Weight: 296.4 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00866994
CAS-Nummer: 107868-30-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 192 to 196 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
HPLC ≥97.5 %
Specific optical rotation +280° to +300° (20 °C, 589 nm) (1% in MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR