Perindopril t-Butylamine Salt

Lieferant: Thermo Scientific

Synonyme: 2-Methylpropan-2-amine (2S,3aS,7aS)-1-((S)-2-(((S)-1-ethoxy-1-oxopentan-2-yl)amino)propanoyl)octahydro-1H-indole-2-carboxylate, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid tert-butylamine salt, Perindopril tert-butylamine, Perindopril erbumine

463210010
ACRO463210010EA 265 EUR
ACRO463210010
Perindopril t-Butylamine Salt
Perindopril t-Butylamine Salt

  • Bioactive

Formel: C₁₉H₃₂N₂O5₅·C₄H₁₁N
Molecular Weight: 441,6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02313824
CAS-Nummer: 107133-36-8

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %
Specific optical rotation -66° to -69° (20°C, 589 nm) (1 % in EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR