Sildenafil citrate
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
5-[2-Ethoxy-5-[(4-methylpiperazin-1-yl)sulphonyl]phenyl]-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one Citrate, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulphonyl]-4-methylpiperazine Citrate
463250010
ACRO463250010EA
104
EUR
ACRO463250010
Sildenafil citrate
Sildenafil citrate
- Bioactive
Formel:
C₂₂H₃₀N₆O₄S·C₆H₈O₇ Molecular Weight: 666,7 g/mol Schmelzpunkt: 182…196 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09026931 CAS-Nummer: 171599-83-0 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 182 to 196 °C |
Loss on drying | ≤2.5 % |
HPLC | ≥97.5 % |
Titration with NaOH | 28.1 to 29.5 % (citric acid dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...