Dofetilide

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane, N-(4-(2-(Methyl(2-(4-(methylsulphonamido)phenoxy)ethyl)amino)ethyl)phenyl)methanesulphonamide

463370010 463372500
ACRO463370010EA 939 EUR
ACRO463370010 ACRO463372500
Dofetilide
Dofetilide

  • Bioactive

Formel: C₁₉H₂₇N₃O₅S₂
Schmelzpunkt: 149…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869707
CAS-Nummer: 115256-11-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 157 °C
Water ≤1 %
Solubility (2 % in DMSO) Clear solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR