Etoricoxib
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
MK0663, L-791456, MK-0663, 5-Chloro-6'-methyl-3-[4-(methylsulphonyl)phenyl]-2,3'-bipyridine, Arcoxia, 5-Chloro-6'-methyl-3-(4-(methylsulphonyl)phenyl)-2,3'-bipyridine
463380010
ACRO463380010EA
121
EUR
ACRO463380010
Etoricoxib
Etoricoxib
- Bioactive
Formel:
C₁₈H₁₅ClN₂O₂S Molecular Weight: 358,84 g/mol Schmelzpunkt: 134…138 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06797512 CAS-Nummer: 202409-33-4 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Crsytalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.1 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...