(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride

Lieferant: Thermo Fisher Scientific

463420010 463420050 463420250
ACRO463420010EA 34.7 EUR
ACRO463420010 ACRO463420050 ACRO463420250
(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride
(6R,7R)-7-((Z)-2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride

  • Bioactive

Formel: C₁₉H₁₈ClN₅O₇S₃
Schmelzpunkt: >190 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 103980-44-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige Powder or crystals
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥97.5 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR