Tinidazole

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-[2-(Ethylsulphonyl)ethyl]-2-methyl-5-nitroimidazole, 1-(2-(Ethylsulphonyl)ethyl)-2-methyl-5-nitro-1H-imidazole

463510050 463510250
ACRO463510050EA 29.8 EUR
ACRO463510050 ACRO463510250
Tinidazole
Tinidazole

  • Bioactive

Formel: C₈H₁₃N₃O₄S
Molecular Weight: 247.27 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057217
CAS-Nummer: 19387-91-8

Bestellen

Spezifikation Testergebnisse

Appearance Almost white to light yellow Powder
Infrared spectrum Conforms
Melting point 125 to 128 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR