Aliskiren hemifumarate

Lieferant: Thermo Fisher Scientific

Synonyme: Rasilez, (2S,4S,5S,7S)-N-(2-Carbamoyl-2-methylpropyl)-5-amino-4-hydroxy-2,7diisopropyl-8-[4-methoxy-3-(3-methoxypropoxy)phenyl]-octanamide hemifumarate, Aliskiren Hemifumarate, SPP-100, CGP-60536, (2S,4S,5S,7S)-N-(2-Carbamoyl-2-methylpropyl)-5-amino-4-hydroxy-2,7diisopropyl-8-[4-methoxy-3-(3-methoxypropoxy)phenyl]-octanamide hemifumarate, CGP60536B, (2S,4S,5S,7S)-5-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-2-isopropyl-7-(4-methoxy-3-(3-methoxypropoxy)benzyl)-8-methylnonanamide hemifumarate

463790010 463792500
ACRO463790010EA 412 EUR
ACRO463790010 ACRO463792500
Aliskiren hemifumarate
Aliskiren hemifumarate

  • Bioactive

Formel: C₃₀H₅₃N₃O₆·0.5C₄H₄O₄
Lagertemperatur: Kühlschrank
MDL: MFCD27756642
CAS-Nummer: 173334-58-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 9.3 to 9.7 % (fumaric acid dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR