Carteolol hydrochloride

Lieferant: Thermo Fisher Scientific
Achtung

464320010 464320050
ACRO464320010EA 109 EUR
ACRO464320010 ACRO464320050
Carteolol hydrochloride
Carteolol hydrochloride

  • Bioactive

Formel: C₁₆H₂₄N₂O₃·HCl
Schmelzpunkt: 278 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00941499
CAS-Nummer: 51781-21-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C)
Solubility (0.3 g in 10 ml water) Clear colorless solution
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR