Benzophenone azine

Lieferant: Thermo Scientific

464580010
ACRO464580010EA 108 EUR
ACRO464580010
Benzophenone azine
Benzophenone azine

  • Bioactive

Formel: C₂₆H₂₀N₂
Molecular Weight: 360,46 g/mol
Schmelzpunkt: 161…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022006
CAS-Nummer: 983-79-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR