Ammoniumchlorid für die Molekularbiologie

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Salmiak

464595000
ACRO464595000EA 109 EUR
ACRO464595000
Ammoniumchlorid für die Molekularbiologie
Ammoniumchlorid

  • Bioactive

Formel: NH₄Cl
Schmelzpunkt: 340 °C (sublimes) (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011420
CAS-Nummer: 12125-02-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder, crystals and/or chunks
Solubility (10 % in water) Clear colorless solution
Titration Argentimetric ≥99.4 %
pH 4.5 to 5.5
Dnase,Rnase,Protease activity none detected
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR