Neratinib

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: HKI-272, (E)-N-(4-((3-Chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide

464700010 464702500
ACRO464700010EA 264 EUR
ACRO464700010 ACRO464702500
Neratinib
Neratinib

  • Bioactive

Formel: C₃₀H₂₉ClN₆O₃
Molecular Weight: 557 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09752958
CAS-Nummer: 698387-09-6

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to beige Powder
Infrared spectrum Conforms
Solubility (1% in DMSO) Clear colorless to light yellow
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR