N-Succinimidyl Acetate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate
464750050
464750250
464751000
ACRO464750050EA
51.7
EUR
ACRO464750050
ACRO464750250
ACRO464751000
N-Succinimidyl Acetate
N-Succinimidyl Acetate
- Bioactive
Formel:
C₆H₇NO₄ Molecular Weight: 157.13 g/mol Schmelzpunkt: 133…134 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037916 CAS-Nummer: 14464-29-0 |
Spezifikation Testergebnisse
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 132 to 136 °C |
Water | ≤2 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...