Bendazac

Lieferant: Thermo Scientific
Achtung

Synonyme: [(1-Benzyl-3-indazolyl)oxy]acetic Acid, Bendazolic acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole

464920010
ACRO464920010EA 52.1 EUR
ACRO464920010
Bendazac
Bendazac
Formel: C₁₆H₁₄N₂O₃
Molecular Weight: 282,30 g/mol
Schmelzpunkt: 160°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00866158
CAS-Nummer: 20187-55-7
EINECS: 243-569-2
Merck Index: 13.1032

Bestellen

Spezifikation Testergebnisse

Appearance White powder
Infrared spectrum Conforms
Melting point 160°C to 164°C
HPLC ≥ 97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR