Zonisamide

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,2-Benzisoxazole-3-methanesulphonamide, Benzo[d]isoxazol-3-ylmethanesulphonamide

465040010 465042500
ACRO465040010EA 205 EUR
ACRO465040010 ACRO465042500
Zonisamide
Zonisamide

  • Bioactive

Formel: C₈H₈N₂O₃S
Schmelzpunkt: 165 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00865316
CAS-Nummer: 68291-97-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
Water ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR