Zonisamide
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,2-Benzisoxazole-3-methanesulphonamide, Benzo[d]isoxazol-3-ylmethanesulphonamide
465040010
465042500
ACRO465040010EA
205
EUR
ACRO465040010
ACRO465042500
Zonisamide
Zonisamide
- Bioactive
Formel:
C₈H₈N₂O₃S Schmelzpunkt: 165 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00865316 CAS-Nummer: 68291-97-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 163 to 167 °C |
Water | ≤1 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...