Hydrocortisone 17-Butyrate

Lieferant: Thermo Scientific
Achtung

Synonyme: 11beta,17,21-Trihydroxypregn-4-ene-3,20-dione 17-Butyrate, Cortisol 17-Butyrate

465050010
ACRO465050010EA 91.8 EUR
ACRO465050010
Hydrocortisone 17-Butyrate
Hydrocortisone 17-Butyrate
Formel: C₂₅H₃₆O₆
Molecular Weight: 432,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00083364
CAS-Nummer: 13609-67-1
EINECS: 237-093-4
Merck Index: 13.4808

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Loss on drying ≤ 1%
HPLC ≥ 96.0%
Specific optical rotation +47° to +54° (20°C, 589nm) (0.1% in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR