Azelnidipine

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-(1-Benzhydrylazetidin-3-yl) 5-isopropyl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

465060010
ACRO465060010EA 56.3 EUR
ACRO465060010
Azelnidipine
Azelnidipine
Formel: C₃₃H₃₄N₄O₆
Molecular Weight: 582,66 g/mol
Schmelzpunkt: 193,3°C…195°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00865803
CAS-Nummer: 123524-52-7

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to yellow powder
Infrared spectrum Conforms
Melting point 120°C to 126°C
HPLC ≥ 97.5%
Loss on drying ≤ 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR