Balsalazide disodium salt dihydrate

Lieferant: Thermo Fisher Scientific

465090010 465090050
ACRO465090010EA 44 EUR
ACRO465090010 ACRO465090050
Balsalazide disodium salt dihydrate

  • Bioactive

Formel: C₁₇H₁₇N₃Na₂O₈
Lagertemperatur: Kühlschrank
MDL: MFCD01631117
CAS-Nummer: 150399-21-6

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-red Crystalline powder
Infrared spectrum Conforms
Water 7 to 10 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR