Tolvaptan
Lieferant: Thermo Scientific
465350010
ACRO465350010EA
661
EUR
ACRO465350010
Tolvaptan
Tolvaptan
Formel:
C₂₆H₂₅ClN₂O₃ Molecular Weight: 448,94 g/mol Schmelzpunkt: 225,9°C Lagertemperatur: Kühlschrank |
MDL:
MFCD09838782 CAS-Nummer: 150683-30-0 |
Spezifikation Testergebnisse
Appearance | White to light brown powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...