Parecoxib sodium
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dynastat, SC-69124A, Sodium ((4-(5-methyl-3-phenylisoxazol-4-yl)phenyl)sulphonyl)(propionyl)amide
465380010
ACRO465380010EA
256
EUR
ACRO465380010
Parecoxib sodium
Parecoxib sodium
Formel:
C₁₉H₁₇N₂NaO₄S Molecular Weight: 392,4 g/mol Schmelzpunkt: 273°C…275°C Lagertemperatur: Kühlschrank |
MDL:
MFCD04972608 CAS-Nummer: 198470-85-8 |
Spezifikation Testergebnisse
Appearance | White to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 98.5% |
Water | ≤ 3% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...