Flumetasone
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
465442500
ACRO465442500EA
135
EUR
ACRO465442500
Flumetasone
Flumetasone
- Bioactive
Formel:
C₂₂H₂₈F₂O₅ Molecular Weight: 410.46 g/mol Schmelzpunkt: >238 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00056464 CAS-Nummer: 2135-17-3 |
Spezifikation Testergebnisse
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Specific optical rotation | +66.0° to +71.0° (20°C, 589 nm) (1 % in Dioxane) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...