Gastrodin

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol

465460010
ACRO465460010EA 253 EUR
ACRO465460010
Gastrodin
Gastrodin
Formel: C₁₃H₁₈O₇
Molecular Weight: 286,28 g/mol
Schmelzpunkt: 169°C…170°C
Lagertemperatur: Kühlschrank
MDL: MFCD00272169
CAS-Nummer: 62499-27-8

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white powder
Infrared spectrum Conforms
Melting point 153°C to 157°C
HPLC ≥ 97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR